Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,1,3-Benzoxadiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. CC09701DA
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC09701DA 1 g
CC09701CB 250 mg
CC09701EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC09701DA Supplier Thermo Scientific Chemicals Supplier No. CC09701DA
Add to Cart

Chemical Identifiers

CAS 19155-88-5
Molecular Formula C7H3N2O3
Molecular Weight (g/mol) 163.11
MDL Number MFCD00276977
InChI Key WZUFYJFTOVGJJT-UHFFFAOYSA-M
Synonym benzo c 1,2,5 oxadiazole-5-carboxylic acid, benzofurazan-5-carboxylic acid, 5-benzofurazancarboxylic acid, benzo c 1,2,5-oxadiazole-5-carboxylic acid, pubchem3995, maybridge1_002587, 4-bromo-2,6-dimethylanisole,, 2,1,3-benzoxadiazole-5-carboxylicacid, benzo 1,2,5 oxadiazole-5-carboxylic acid, benzo 2,1,3 oxadiazole-5-carboxylic acid
PubChem CID 2735454
IUPAC Name 2,1,3-benzoxadiazole-5-carboxylic acid
SMILES [O-]C(=O)C1=CC2=NON=C2C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 164.12
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.