Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,1,3-Benzoxadiazole, 97%, Thermo Scientific™

Catalog No. KM06477EA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KM06477EA 10 g
KM06477DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. KM06477EA Supplier Thermo Scientific Chemicals Supplier No. KM06477EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 273-09-6
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.111
InChI Key AWBOSXFRPFZLOP-UHFFFAOYSA-N
Synonym benzofurazan, benzo c 1,2,5 oxadiazole, benzofurazane, 2,1,3-benzoxadiazol, benzo-2,1,3-oxadiazole, 2-oxa-1,3-diaza-2h-isoindene, 3,5-cyclohexadiene-1,2-dione, dioxime, furazan, pubchem22999, acmc-209gwb
PubChem CID 67501
IUPAC Name 2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C=C1

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazole
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.