Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium Bromide 98%, Thermo Scientific™

Catalog No. AC204650050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC204650050 5 g
AC204650250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC204650050 Supplier Thermo Scientific Chemicals Supplier No. 204650050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69891-92-5
Molecular Formula C24H26BrO2P
Molecular Weight (g/mol) 457.35
MDL Number MFCD00012000
InChI Key XETDBHNHTOJWPZ-UHFFFAOYSA-M
Synonym 2-1,3-dioxan-2-yl ethyltriphenylphosphonium bromide, 2-1,3-dioxan-2-yl ethyl triphenylphosphonium bromide, 2-1,3-dioxan-2-yl ethyl triphenylphosphanium bromide, acmc-209obj, xqaupnaycikwct-uhfffaoysa-m, 1,3-dioxan-2-ylethyltriphenylphosphinebromide, 2-1,3-dioxan-2-yl ethyltriphenylphosphonium bromi, 2-1,3-dioxan-2-yl ethyltriphenylphosphoniumbromide, 2-1,3-dioxane-2-yl ethyltriphenylphosphonium bromide
PubChem CID 2724168
IUPAC Name [2-(1,3-dioxan-2-yl)ethyl]triphenylphosphanium bromide
SMILES [Br-].C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1OCCCO1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Brown
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 457.34
Percent Purity ≥97.5% (on anhydrous substance)
Physical Form Powder
Chemical Name or Material [2-(1, 3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide, 98%
Hazard Category HYGROSCOPIC
EINECSNumber 274-189-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.