Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1,3-diphenyl-1H-pyrazol-4-yl)-2,3-dihydroquinazolin-4(1H)-one | 474379-42-5 | 95.0% | 366.42 | C23H18N4O | 10 MG
SDP

Catalog No. 5000286347
Encompass_Preferred

Antioxidant agent-11 (compound 3d) is a small-molecule research compound reported to have weak antioxidant and anticancer activity. It is supplied as a solid for laboratory use and is intended for in vitro research applications; handle with standard laboratory safety procedures and store protected from light.

  • Small-molecule antioxidant with weak anticancer activity.
  • Molecular formula C23H18N4O.
  • Molecular weight 366.42.
  • CAS number 474379-42-5.
  • Available in small milligram pack sizes suitable for screening and assay work.
  • Recommended storage: 4°C and protected from light; in solvent store at -80°C.

Catalog No. 50-002-86347 Supplier Medchemexpress LLC Supplier No. HY15375710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antioxidant agent-11 (compound 3d) is a small-molecule research compound reported to have weak antioxidant and anticancer activity. It is supplied as a solid for laboratory use and is intended for in vitro research applications; handle with standard laboratory safety procedures and store protected from light.

  • Small-molecule antioxidant with weak anticancer activity.
  • Molecular formula C23H18N4O.
  • Molecular weight 366.42.
  • CAS number 474379-42-5.
  • Available in small milligram pack sizes suitable for screening and assay work.
  • Recommended storage: 4°C and protected from light; in solvent store at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.