Learn More
Medchemexpress LLC 2-[1,4,7-triazacyclononan-1-yl-4,7-bis(tBu-ester)]-1,5-pentanedioic acid | 1190101-34-8 | MFCD29083820 | 99.8% | 543.69 g/mol | C27H49N3O8 | 25 MG
SDP
Catalog No. 5000284202
Encompass_Preferred
NODAGA-tris(t-Bu ester) (NODAG) is a bifunctional chelator designed to conjugate peptides, antibodies, and other biomolecules for subsequent radiolabeling in PET imaging. Provided as a solid reagent for research use, it combines high purity with practical solubility and storage properties for laboratory workflows.
  • High purity suitable for conjugation and radiolabeling.
  • Off-white to light yellow solid for easy handling.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Example in vivo formulations achieve ≥2.5 mg/mL clear solutions.
  • Stable under recommended storage: powder at -20°C (3 years) or 4°C (2 years).
  • Suitable for labeling peptides and antibodies for PET imaging.
Catalog No. 50-002-84202 Supplier Medchemexpress LLC Supplier No. HYW53989325MG
Add to Cart
Add to Cart
NODAGA-tris(t-Bu ester) (NODAG) is a bifunctional chelator designed to conjugate peptides, antibodies, and other biomolecules for subsequent radiolabeling in PET imaging. Provided as a solid reagent for research use, it combines high purity with practical solubility and storage properties for laboratory workflows.
  • High purity suitable for conjugation and radiolabeling.
  • Off-white to light yellow solid for easy handling.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Example in vivo formulations achieve ≥2.5 mg/mL clear solutions.
  • Stable under recommended storage: powder at -20°C (3 years) or 4°C (2 years).
  • Suitable for labeling peptides and antibodies for PET imaging.