Learn More
Medchemexpress LLC 2-[1-(4-chloro-phenyl)-cyclopropyl]-7-iodo-5-oxo-4,5-dihydro-pyrazolo[1,5-a]quinazoline-3-carbonitrile | 2226517-75-3 | 98.2% | C20H12ClIN4O | 10MG
SDP
Catalog No. 5000261054
Encompass_Preferred
Dynapyrazole-A is a selective small-molecule inhibitor that targets the ATPase activity of microtubule-stimulated dynamin, used in biochemical and cellular studies to probe dynamin-dependent membrane trafficking and endocytosis processes.
  • Selective inhibitor of microtubule-stimulated dynamin ATPase activity.
  • High purity 98.2% suitable for research assays.
  • Soluble in DMSO (25 mg/mL); available as 10 mM solution in DMSO.
  • Stable as powder at -20°C for extended storage; store solutions at -80°C for best stability.
  • Suitable for biochemical and cell biology studies involving dynamin-dependent processes.
Catalog No. 50-002-61054 Supplier Medchemexpress LLC Supplier No. HY13187310MG
Dynapyrazole-A is a selective small-molecule inhibitor that targets the ATPase activity of microtubule-stimulated dynamin, used in biochemical and cellular studies to probe dynamin-dependent membrane trafficking and endocytosis processes.
  • Selective inhibitor of microtubule-stimulated dynamin ATPase activity.
  • High purity 98.2% suitable for research assays.
  • Soluble in DMSO (25 mg/mL); available as 10 mM solution in DMSO.
  • Stable as powder at -20°C for extended storage; store solutions at -80°C for best stability.
  • Suitable for biochemical and cell biology studies involving dynamin-dependent processes.