Learn More
SKYLINE CENTER Item #W393 Bag 2 1/4” Round - 500ct.
Catalog No. NC9864348
Encompass
2 1/4” ROUND 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - .35 Rifle - 20 Gauge
Catalog No. NC9864348 Supplier SKYLINE CENTER Supplier No. 393
2 1/4” ROUND 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - .35 Rifle - 20 Gauge