Learn More
Cayman Chemical 2-(1,8-Naphthyridin-2-yl)phenol, Purity: ≥98%, CAS: 65182-56-1, 10 mg
SDP
Catalog No. 502991802
Encompass_Preferred
A STAT1 activity enhancer; enhances IFN-γ-induced STAT1-dependent signaling at 45 µM; increases IFN-γ-induced IRF3 expression and STAT1 phosphorylation in NIH3T3 cells at 45 µM; increases growth inhibition of MCF-7 and 2fTGH cancer cells induced by IFN-γ at 45 µM
Catalog No. 50-299-1802 Supplier Cayman Chemical Supplier No. 3091810 MG
A STAT1 activity enhancer; enhances IFN-γ-induced STAT1-dependent signaling at 45 µM; increases IFN-γ-induced IRF3 expression and STAT1 phosphorylation in NIH3T3 cells at 45 µM; increases growth inhibition of MCF-7 and 2fTGH cancer cells induced by IFN-γ at 45 µM