Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-benzylpiperidin-4-yl)thiochroman-4-one | 1204401-49-9 | 99.9% | 337.48 g·mol⁻¹ | C21H23NOS | 100 MG
SDP

Catalog No. 5000243086
Encompass_Preferred

σ1 Receptor antagonist used as a selective sigma-1 receptor antagonist for pharmacological and cell-based research. Supplied as a solid powder for laboratory use.

  • Selective sigma-1 receptor antagonist suitable for in vitro studies.
  • High purity of 99.9% for reliable experimental results.
  • Chemical formula C21H23NOS and molecular weight 337.48 g·mol⁻¹.
  • Soluble in DMSO at 100 mg/mL (may require sonication).
  • Appearance: solid, light yellow to light brown.
  • Stable when stored as powder at -20°C for up to 3 years.
  • Available in small research quantities (100 MG) for laboratory use.

Catalog No. 50-002-43086 Supplier Medchemexpress LLC Supplier No. HY10815100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

σ1 Receptor antagonist used as a selective sigma-1 receptor antagonist for pharmacological and cell-based research. Supplied as a solid powder for laboratory use.

  • Selective sigma-1 receptor antagonist suitable for in vitro studies.
  • High purity of 99.9% for reliable experimental results.
  • Chemical formula C21H23NOS and molecular weight 337.48 g·mol⁻¹.
  • Soluble in DMSO at 100 mg/mL (may require sonication).
  • Appearance: solid, light yellow to light brown.
  • Stable when stored as powder at -20°C for up to 3 years.
  • Available in small research quantities (100 MG) for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.