Learn More
Medchemexpress LLC 2-[[1-(cyclopropylmethoxy)-4-hydroxy-2-oxoquinoline-3-carbonyl]amino]acetic acid | 1616690-16-4 | 99.1% | 332.31 g/mol | C16H16N2O6 | 25 MG
SDP
Catalog No. 5000247260
Encompass_Preferred
Desidustat is an orally active hypoxia-inducible factor (HIF) prolyl hydroxylase inhibitor used in research to stimulate erythropoiesis and to study anemia and ischemia/hypoxia-related conditions. The compound is supplied in solid and solution formats and is characterized by high purity and defined molecular properties.
  • Orally active HIF prolyl hydroxylase inhibitor for research use.
  • Used to model anemia and hypoxia/ischemia-related conditions.
  • High reported purity (99.1%).
  • Molecular formula C16H16N2O6; molecular weight 332.31 g/mol.
  • Available as small solid packs and DMSO solutions for assay preparation.
Catalog No. 50-002-47260 Supplier Medchemexpress LLC Supplier No. HY10322725MG
Desidustat is an orally active hypoxia-inducible factor (HIF) prolyl hydroxylase inhibitor used in research to stimulate erythropoiesis and to study anemia and ischemia/hypoxia-related conditions. The compound is supplied in solid and solution formats and is characterized by high purity and defined molecular properties.
  • Orally active HIF prolyl hydroxylase inhibitor for research use.
  • Used to model anemia and hypoxia/ischemia-related conditions.
  • High reported purity (99.1%).
  • Molecular formula C16H16N2O6; molecular weight 332.31 g/mol.
  • Available as small solid packs and DMSO solutions for assay preparation.