Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione 98.0+%, TCI America™
SDP

Catalog No. H15465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15465G 5 g
H15461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15465G Supplier TCI America Supplier No. H15465G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 172611-72-2
Molecular Formula C13H20O3
Molecular Weight (g/mol) 224.3
MDL Number MFCD01862890
InChI Key HNPWTDUZIXAJSA-UHFFFAOYSA-N
Synonym 2-(1-Hydroxy-3-methylbutylidene)dimedone
PubChem CID 22728397
IUPAC Name 5,5-dimethyl-2-(3-methylbutanoyl)cyclohexane-1,3-dione
SMILES CC(C)CC(=O)C1C(=O)CC(CC1=O)(C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 224.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(1-Hydroxy-3-methylbutylidene)-5,5-dimethyl-1,3-cyclohexanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.