Learn More
Medchemexpress LLC HY-P1868A 1mg Medchemexpress, α2β1 Integrin Ligand Peptide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1868A1MG
Encompass_Preferred
Medchemexpress, HY-P1868A 1mg α2β1 Integrin Ligand Peptide (TFA) CAS: α2β1 Integrin Ligand Peptide TFA interacts with the α2β1 integrin receptor on the cell membrane and mediates extracellular signals into cells. It is a potential antagonist of collagen receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0543