Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-methylethyl)phenyl diphenyl phosphate | 64532-94-1 | MFCD00870203 | 368.36 | C21H21O4P | 25 MG
SDP

Catalog No. 5000284583
Encompass_Preferred

2-(1-Methylethyl)phenyl diphenyl phosphate is an aryl diphenyl phosphate compound primarily used as a flame retardant in research and material testing. It is an aromatic phosphate ester with CAS number 64532-94-1, molecular formula C21H21O4P, and molecular weight 368.36.

  • Aryl diphenyl phosphate used as a flame retardant.
  • CAS number 64532-94-1 for unambiguous identification.
  • Molecular formula C21H21O4P; molecular weight 368.36.
  • Suitable for analytical and research applications at small sample sizes.

Catalog No. 50-002-84583 Supplier Medchemexpress LLC Supplier No. HYW58826725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(1-Methylethyl)phenyl diphenyl phosphate is an aryl diphenyl phosphate compound primarily used as a flame retardant in research and material testing. It is an aromatic phosphate ester with CAS number 64532-94-1, molecular formula C21H21O4P, and molecular weight 368.36.

  • Aryl diphenyl phosphate used as a flame retardant.
  • CAS number 64532-94-1 for unambiguous identification.
  • Molecular formula C21H21O4P; molecular weight 368.36.
  • Suitable for analytical and research applications at small sample sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.