Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-methylethyl)phenyl diphenyl phosphate | 64532-94-1 | MFCD00870203 | 368.36 g·mol⁻¹ | C21H21O4P | 1 G
SDP

Catalog No. 5000284409
Encompass_Preferred

2-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for research use. It is used as a flame retardant additive in materials and as a reference standard for analytical and toxicological studies. Not for human use.

  • Organophosphate flame retardant for research use
  • Chemical formula: C21H21O4P
  • Molecular weight: 368.36 g·mol⁻¹
  • CAS number: 64532-94-1
  • Suitable for material flame-retardancy studies and analytical reference work

Catalog No. 50-002-84409 Supplier Medchemexpress LLC Supplier No. HYW5882671G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for research use. It is used as a flame retardant additive in materials and as a reference standard for analytical and toxicological studies. Not for human use.

  • Organophosphate flame retardant for research use
  • Chemical formula: C21H21O4P
  • Molecular weight: 368.36 g·mol⁻¹
  • CAS number: 64532-94-1
  • Suitable for material flame-retardancy studies and analytical reference work

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.