Learn More
Medchemexpress LLC 2-(1-methylethyl)phenyl diphenyl phosphate | 64532-94-1 | MFCD00870203 | 95.0% | 368.36 g/mol | C21H21O4P | 250 MG
SDP
Catalog No. 5000284275
Encompass_Preferred
2-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for laboratory research use. It serves as a flame retardant additive and analytical reference standard; the compound has formula C21H21O4P, molecular weight 368.36 g/mol, and CAS 64532-94-1.
  • Organophosphate flame retardant for materials and analytical research.
  • Molecular formula C21H21O4P; molecular weight 368.36 g/mol.
  • Available in small laboratory quantities (25 MG-1 G) for analytical workflows.
  • Product datasheet and documentation available; confirm purity from the certificate of analysis.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-84275 Supplier Medchemexpress LLC Supplier No. HYW588267250MG
2-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for laboratory research use. It serves as a flame retardant additive and analytical reference standard; the compound has formula C21H21O4P, molecular weight 368.36 g/mol, and CAS 64532-94-1.
  • Organophosphate flame retardant for materials and analytical research.
  • Molecular formula C21H21O4P; molecular weight 368.36 g/mol.
  • Available in small laboratory quantities (25 MG-1 G) for analytical workflows.
  • Product datasheet and documentation available; confirm purity from the certificate of analysis.
  • For research use only; not for human or clinical use.