Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-methylethyl)phenyl diphenyl phosphate | 64532-94-1 | MFCD00870203 | 97.0% | 368.36 g/mol | C21H21O4P | 50 MG
SDP

Catalog No. 5000284028
Encompass_Preferred

2-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for research use. It is employed as an additive in materials and polymer studies to assess flame retardancy and thermal behavior.

  • Organophosphate flame retardant for materials research.
  • CAS number 64532-94-1 and molecular weight 368.36 g/mol.
  • Molecular formula C21H21O4P.
  • Available in small pack sizes suitable for laboratory testing (25 mg-10 g).
  • Lot-specific purity reported on the Certificate of Analysis; refer to CoA for exact value.
  • Refer to the safety data sheet for handling and storage requirements.

Catalog No. 50-002-84028 Supplier Medchemexpress LLC Supplier No. HYW58826750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(1-Methylethyl)phenyl diphenyl phosphate is an organophosphate flame retardant supplied for research use. It is employed as an additive in materials and polymer studies to assess flame retardancy and thermal behavior.

  • Organophosphate flame retardant for materials research.
  • CAS number 64532-94-1 and molecular weight 368.36 g/mol.
  • Molecular formula C21H21O4P.
  • Available in small pack sizes suitable for laboratory testing (25 mg-10 g).
  • Lot-specific purity reported on the Certificate of Analysis; refer to CoA for exact value.
  • Refer to the safety data sheet for handling and storage requirements.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.