Learn More
Medchemexpress LLC 2-(1-octynyl)adenosine | 90596-75-1 | 375.42 g·mol⁻¹ | C18H25N5O4 | 1 MG
SDP
Catalog No. 5000243578
Encompass_Preferred
YT 146 is a potent adenosine A2 receptor (A2AR) agonist used in pharmacological research. It induces concentration-dependent cAMP accumulation (EC50 ≈ 1.5 nM) and has reported cardioprotective effects, making it suitable for studies of adenosine signaling and cardiovascular pharmacology.
  • Potent A2 adenosine receptor agonist (EC50 ≈ 1.5 nM).
  • Causes concentration-dependent cAMP accumulation.
  • Applicable to cardiovascular and receptor signaling studies.
  • Molecular formula C18H25N5O4; molecular weight 375.42 g·mol⁻¹.
  • Use appropriate laboratory safety and handling procedures.
Catalog No. 50-002-43578 Supplier Medchemexpress LLC Supplier No. HY1063031MG
YT 146 is a potent adenosine A2 receptor (A2AR) agonist used in pharmacological research. It induces concentration-dependent cAMP accumulation (EC50 ≈ 1.5 nM) and has reported cardioprotective effects, making it suitable for studies of adenosine signaling and cardiovascular pharmacology.
  • Potent A2 adenosine receptor agonist (EC50 ≈ 1.5 nM).
  • Causes concentration-dependent cAMP accumulation.
  • Applicable to cardiovascular and receptor signaling studies.
  • Molecular formula C18H25N5O4; molecular weight 375.42 g·mol⁻¹.
  • Use appropriate laboratory safety and handling procedures.