Learn More
Medchemexpress LLC 2-(1-phenyl-cyclopropylamino)-pyrimidine-5-carboxylic acid ethyl ester | 1598426-03-9 | MFCD28347705 | 98.1% | 283.33 g·mol⁻¹ | C16H17N3O2 | 5 MG
SDP
Catalog No. 5000222028
Encompass_Preferred
CG347B is a selective histone deacetylase 6 (HDAC6) inhibitor used as a research tool in oncology, immunology, and neurology studies and as an intermediate in the synthesis of metalloenzyme inhibitors. It is supplied as a white to off-white solid (C16H17N3O2, 283.33 g·mol⁻¹) with reported high purity and well-characterized solubility properties for in vitro and in vivo formulations.
  • Selective HDAC6 inhibition for epigenetics research
  • High reported purity (~98.1%) for reliable experimental results
  • Solid, white to off-white form for straightforward handling and storage
  • High DMSO solubility (250 mg/mL) suitable for in vitro assays
  • Soluble in in vivo vehicle at ≥6.25 mg/mL for formulation studies
  • Available in small research quantities for screening and pilot studies
  • Manufacturer-provided datasheet, SDS, and COA available for quality assurance
Catalog No. 50-002-22028 Supplier Medchemexpress LLC Supplier No. HY1358905MG
CG347B is a selective histone deacetylase 6 (HDAC6) inhibitor used as a research tool in oncology, immunology, and neurology studies and as an intermediate in the synthesis of metalloenzyme inhibitors. It is supplied as a white to off-white solid (C16H17N3O2, 283.33 g·mol⁻¹) with reported high purity and well-characterized solubility properties for in vitro and in vivo formulations.
  • Selective HDAC6 inhibition for epigenetics research
  • High reported purity (~98.1%) for reliable experimental results
  • Solid, white to off-white form for straightforward handling and storage
  • High DMSO solubility (250 mg/mL) suitable for in vitro assays
  • Soluble in in vivo vehicle at ≥6.25 mg/mL for formulation studies
  • Available in small research quantities for screening and pilot studies
  • Manufacturer-provided datasheet, SDS, and COA available for quality assurance