Learn More
Medchemexpress LLC 2-(1-Piperazinyl)pyrimidine | 20980-22-7 | 164.21 | 1 G
SDP
Catalog No. 5000446272
Encompass_Preferred
2-(1-Piperazinyl)pyrimidine is the major metabolite of Tandospirone (HY-14558), categorized as an Endogenous Metabolite with a Metabolic Enzyme/Protease pathway. This product is for research use only and has not been fully validated for medical applications.
  • Major metabolite of Tandospirone
  • Categorized as an endogenous metabolite
  • Appears as colorless to off-white solid below 32°C and liquid above 34°C
  • High purity of 99.94%
  • Soluble in DMSO at 100 mg/mL
  • Structure confirmed by 1H NMR and LCMS
Catalog No. 50-004-46272 Supplier Medchemexpress LLC Supplier No. HYW0044641G
2-(1-Piperazinyl)pyrimidine is the major metabolite of Tandospirone (HY-14558), categorized as an Endogenous Metabolite with a Metabolic Enzyme/Protease pathway. This product is for research use only and has not been fully validated for medical applications.
  • Major metabolite of Tandospirone
  • Categorized as an endogenous metabolite
  • Appears as colorless to off-white solid below 32°C and liquid above 34°C
  • High purity of 99.94%
  • Soluble in DMSO at 100 mg/mL
  • Structure confirmed by 1H NMR and LCMS