Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-Piperazinyl)pyrimidine | 20980-22-7 | 164.21 | 5 G
SDP

Catalog No. 5000449231
Encompass_Preferred

2-(1-Piperazinyl)pyrimidine is identified as the major metabolite of Tandospirone and is intended for research use only. It appears as a colorless to off-white solid below 32°C and as a liquid above 34°C.

  • Major metabolite of Tandospirone
  • For research use only
  • Colorless to off-white appearance
  • Solid below 32°C, liquid above 34°C
  • Stable in pure form at -20°C for 3 years and 4°C for 2 years
  • Stable in solvent at -80°C for 6 months and -20°C for 1 month

Catalog No. 50-004-49231 Supplier Medchemexpress LLC Supplier No. HYW0044645G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(1-Piperazinyl)pyrimidine is identified as the major metabolite of Tandospirone and is intended for research use only. It appears as a colorless to off-white solid below 32°C and as a liquid above 34°C.

  • Major metabolite of Tandospirone
  • For research use only
  • Colorless to off-white appearance
  • Solid below 32°C, liquid above 34°C
  • Stable in pure form at -20°C for 3 years and 4°C for 2 years
  • Stable in solvent at -80°C for 6 months and -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.