Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1-Piperazinyl)pyrimidine | 20980-22-7 | 164.21 | 500 UG
SDP

Catalog No. 5000448823
Encompass_Preferred

2-(1-Piperazinyl)pyrimidine is the major metabolite of Tandospirone (HY-14558). This high-purity compound has a molecular weight of 164.21 and appears as a colorless to off-white solid or liquid depending on temperature.

  • Major metabolite of Tandospirone
  • Purity: 99.94%
  • Molecular formula: C8H12N4
  • Molecular weight: 164.21
  • Appearance: Colorless to off-white solid or liquid
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-48823 Supplier Medchemexpress LLC Supplier No. HYW004464S500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-(1-Piperazinyl)pyrimidine is the major metabolite of Tandospirone (HY-14558). This high-purity compound has a molecular weight of 164.21 and appears as a colorless to off-white solid or liquid depending on temperature.

  • Major metabolite of Tandospirone
  • Purity: 99.94%
  • Molecular formula: C8H12N4
  • Molecular weight: 164.21
  • Appearance: Colorless to off-white solid or liquid
  • Store pure form at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.