Learn More
Medchemexpress LLC 2-(1-Piperazinyl)pyrimidine (Standard) | 20980-22-7 | 164.21 | 100 MG
SDP
Catalog No. 5000445472
Encompass_Preferred
2-(1-Piperazinyl)pyrimidine (Standard) is the analytical standard of 2-(1-Piperazinyl)pyrimidine. This product is intended for research and analytical applications. It is the major metabolite of Tandospirone.
  • Used as a reference standard for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-45472 Supplier Medchemexpress LLC Supplier No. HYW004464R100MG
2-(1-Piperazinyl)pyrimidine (Standard) is the analytical standard of 2-(1-Piperazinyl)pyrimidine. This product is intended for research and analytical applications. It is the major metabolite of Tandospirone.
  • Used as a reference standard for assays
  • Commonly utilized in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications