Learn More
Medchemexpress LLC 2-(1-piperazinyl)pyrimidine (standard) | 20980-22-7 | 164.21 | 25 MG
SDP
Catalog No. 5000448370
Encompass_Preferred
2-(1-Piperazinyl)pyrimidine (Standard) is an analytical standard and the major metabolite of Tandospirone. Intended for research and analytical applications, this compound is suitable for qualitative, quantitative, and methodological research experiments.
  • Analytical standard
  • Reference standard supplied assay
  • Used in HPLC, GC, and MS research
  • Suitable for qualitative, quantitative, and methodological research experiments
Catalog No. 50-004-48370 Supplier Medchemexpress LLC Supplier No. 25MGHYW004464R
2-(1-Piperazinyl)pyrimidine (Standard) is an analytical standard and the major metabolite of Tandospirone. Intended for research and analytical applications, this compound is suitable for qualitative, quantitative, and methodological research experiments.
  • Analytical standard
  • Reference standard supplied assay
  • Used in HPLC, GC, and MS research
  • Suitable for qualitative, quantitative, and methodological research experiments