Learn More
Medchemexpress LLC 2-(1-Piperazinyl)pyrimidine (Standard) | 20980-22-7 | 164.21 | 50 MG
SDP
Catalog No. 5000448903
Encompass_Preferred
2-(1-Piperazinyl)pyrimidine is an analytical standard for 2-(1-Piperazinyl)pyrimidine, suitable for research and analytical applications. It serves as the primary metabolite of Tandospirone (HY-14558) and is utilized as a reference standard in various research experiments, including qualitative, quantitative, and methodological studies such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Primary metabolite
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS
Catalog No. 50-004-48903 Supplier Medchemexpress LLC Supplier No. HYW004464R50MG
2-(1-Piperazinyl)pyrimidine is an analytical standard for 2-(1-Piperazinyl)pyrimidine, suitable for research and analytical applications. It serves as the primary metabolite of Tandospirone (HY-14558) and is utilized as a reference standard in various research experiments, including qualitative, quantitative, and methodological studies such as HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Primary metabolite
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
  • Suitable for techniques such as HPLC, GC, and MS