Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Piperazinylcarbonyl)-1,4-benzodioxane 95.0+%, TCI America™
SDP

Catalog No. P18365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18365G Supplier TCI America Supplier No. P18365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 70918-00-2
Molecular Formula C13H16N2O3
Molecular Weight (g/mol) 248.282
MDL Number MFCD01837119
InChI Key FLUPDJNTYCSBJZ-UHFFFAOYSA-N
Synonym 1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine
PubChem CID 2735449
IUPAC Name 2,3-dihydro-1,4-benzodioxin-3-yl(piperazin-1-yl)methanone
SMILES C1CN(CCN1)C(=O)C2COC3=CC=CC=C3O2

Specifications

Melting Point 105°C
Color Yellow
Quantity 5 g
Formula Weight 248.28
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.