Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1-Pyrrolidinyl)nicotinic acid, 95%, Thermo Scientific™

Catalog No. MO01300DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO01300DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01300DE Supplier Thermo Scientific Chemicals Supplier No. MO01300DE
Add to Cart

Chemical Identifiers

CAS 690632-36-1
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD05865128
InChI Key DTUSJCJZQJAPGD-UHFFFAOYSA-N
Synonym 2-1-pyrrolidinyl nicotinic acid, 2-pyrrolidin-1-yl pyridine-3-carboxylic acid, 2-pyrrolidin-1-ylnicotinic acid, 2-pyrrolidin-1-yl nicotinic acid, 2-pyrrolidinylpyridine-3-carboxylic acid, 2-1-pyrrolidinyl nicotinicacid, 3-pyridinecarboxylic acid, 2-1-pyrrolidinyl
PubChem CID 2794739
SMILES OC(=O)C1=C(N=CC=C1)N1CCCC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 5 g
IUPAC Name 2-pyrrolidin-1-ylpyridine-3-carboxylic acid
Formula Weight 192.22
Percent Purity 95%
Chemical Name or Material 2-(1-Pyrrolidinyl)nicotinic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.