Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(1-pyrrolyl)benzylamine

Catalog No. CDS014675 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014675 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014675 Supplier Sigma Aldrich Supplier No. CDS014675100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Various 3-pyrrolines (2,5-dihydropyrroles) have been synthesized by two-step reaction sequence of alkylation/alkylidene carbene ch-insertion reaction
  • Synthesis of 3-pyrroline has been reported by employing cis-1,4-dichloro-2-butene as starting reagent.3-pyrroline is suitable for use in a study to investigate the core-level binding energies of simple unsaturated organic molecules bonded to the si(001) surface by x-ray photoelectron spectroscopy (xps)
  • It may be used in the synthesis of renin inhibitors and vasodilators

Specifications

Molecular Formula C11H12N2
Linear Formula C11H12N2
MDL Number MFCD03763920
Quantity 1 g
Molecular Weight (g/mol) 172.23
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.