Learn More
Waters Corp WATERS CORPORATION Cartridge Column Holder, 2.1 mm, 20 mm
Catalog No. 50819165 Shop All Waters Corporation Products
Encompass_Preferred
This Cartridge Column Holder is designed to securely hold cartridge columns measuring 2.1 mm x 20 mm. It integrates simply into existing liquid chromatography systems without requiring external utilities, which can improve lab efficiency and reduce overall running expenses.
  • Designed to hold and fit cartridge columns with a length of 2.1 mm x 20 mm.
  • Available in various versions to accommodate different cartridge columns based on preparative chromatographic needs.
  • Simple to integrate into an existing liquid chromatography system.
  • Requires no external utilities.
  • No end-fittings for columns to connect or reconnect.
  • Manufactured in dedicated facilities to high industry standards, including cGMP and ISO 9001:2000, using ultra-pure reagents.
  • Each batch of packaging material and equipment is tested to ensure high-quality materials and consistent, repeatable performance.
Catalog No. 50-819-165 Supplier Waters Corp Supplier No. 186000262
This Cartridge Column Holder is designed to securely hold cartridge columns measuring 2.1 mm x 20 mm. It integrates simply into existing liquid chromatography systems without requiring external utilities, which can improve lab efficiency and reduce overall running expenses.
  • Designed to hold and fit cartridge columns with a length of 2.1 mm x 20 mm.
  • Available in various versions to accommodate different cartridge columns based on preparative chromatographic needs.
  • Simple to integrate into an existing liquid chromatography system.
  • Requires no external utilities.
  • No end-fittings for columns to connect or reconnect.
  • Manufactured in dedicated facilities to high industry standards, including cGMP and ISO 9001:2000, using ultra-pure reagents.
  • Each batch of packaging material and equipment is tested to ensure high-quality materials and consistent, repeatable performance.