Learn More
Medchemexpress LLC 2-((1E,3Z)-3-(1-(1-((E)-Cyclooct-2-en-1-yloxy)-1,12-dioxo-5,8-dioxa-2,11-diazaheptadecan-17-yl)-3,3- | 91.1% | 775.46 | C45H63ClN4O5 | 10 MG
SDP
Catalog No. 5000287486
Encompass_Preferred
Cy3-PEG2-TCO is a Cy3-based fluorescent reagent containing a two-unit PEG spacer and a trans-cyclooctene (TCO) functional group. It is designed for rapid bioorthogonal conjugation via inverse electron-demand Diels-Alder (iEDDA) reactions with tetrazine-bearing partners and is intended for research use in labeling, imaging, and bioconjugation workflows.
  • Fluorescent Cy3 dye derivative suitable for imaging and labeling.
  • PEG2 spacer improves solubility and reduces steric hindrance during conjugation.
  • TCO functional group enables fast, selective iEDDA click reactions with tetrazines.
  • Supplied as a solid with color ranging brown to reddish brown.
  • High purity (~91.1%) with molecular weight 775.46 g·mol-1.
  • Store protected from light under nitrogen at -20°C; in solvent, store at -80°C for long term.
Catalog No. 50-002-87486 Supplier Medchemexpress LLC Supplier No. HY15532210MG
Cy3-PEG2-TCO is a Cy3-based fluorescent reagent containing a two-unit PEG spacer and a trans-cyclooctene (TCO) functional group. It is designed for rapid bioorthogonal conjugation via inverse electron-demand Diels-Alder (iEDDA) reactions with tetrazine-bearing partners and is intended for research use in labeling, imaging, and bioconjugation workflows.
  • Fluorescent Cy3 dye derivative suitable for imaging and labeling.
  • PEG2 spacer improves solubility and reduces steric hindrance during conjugation.
  • TCO functional group enables fast, selective iEDDA click reactions with tetrazines.
  • Supplied as a solid with color ranging brown to reddish brown.
  • High purity (~91.1%) with molecular weight 775.46 g·mol-1.
  • Store protected from light under nitrogen at -20°C; in solvent, store at -80°C for long term.