Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™

Catalog No. cc47501da
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC47501DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC47501DA Supplier Thermo Scientific Chemicals Supplier No. CC47501DA
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 876718-01-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H9N3O2
Quantity 1 g
MDL Number MFCD07186455
Synonym 2-1h-1,2,4-triazol-1-ylmethyl benzoic acid, 2-1,2,4 triazol-1-ylmethyl-benzoic acid, 2-1,2,4-triazol-1-ylmethyl benzoic acid, 2-1,2,4-triazolylmethyl benzoic acid, 2-1h-1,2,4-triazol-1-yl methyl benzoic acid
InChI Key KPKNLIVZZRDKGI-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=C1CN1C=NC=N1
IUPAC Name 2-[(1H-1,2,4-triazol-1-yl)methyl]benzoic acid
Molecular Weight (g/mol) 203.20
PubChem CID 3159712
Formula Weight 203.2
Percent Purity 97%
Chemical Name or Material 2-(1H-1, 2, 4-Triazol-1-ylmethyl)benzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.