Learn More
Medchemexpress LLC 2-[(1H-benzotriazol-6-ylamino)methyl]-4,6-dichlorophenol | 866927-10-8 | ≥99.0% | 309.15 | C13H10Cl2N4O | 25 MG
SDP
Catalog No. 5000213913
Encompass_Preferred
IC87201 is a small-molecule inhibitor of the protein-protein interaction between postsynaptic density protein 95 (PSD95) and neuronal nitric oxide synthase (nNOS). It suppresses NMDA receptor-dependent nitric oxide (NO) and cyclic GMP (cGMP) formation and is used as a research reagent to probe PSD95-nNOS-mediated neuronal signaling.
  • Inhibits PSD95-nNOS protein-protein interaction.
  • Reduces NMDAR-dependent NO and cGMP formation.
  • Useful for studying neuronal signaling and neuroprotection mechanisms.
  • Available in solid and DMSO solution formats for flexible dosing.
  • High purity suitable for biochemical and cellular assays.
Catalog No. 50-002-13913 Supplier Medchemexpress LLC Supplier No. HY10045725MG
IC87201 is a small-molecule inhibitor of the protein-protein interaction between postsynaptic density protein 95 (PSD95) and neuronal nitric oxide synthase (nNOS). It suppresses NMDA receptor-dependent nitric oxide (NO) and cyclic GMP (cGMP) formation and is used as a research reagent to probe PSD95-nNOS-mediated neuronal signaling.
  • Inhibits PSD95-nNOS protein-protein interaction.
  • Reduces NMDAR-dependent NO and cGMP formation.
  • Useful for studying neuronal signaling and neuroprotection mechanisms.
  • Available in solid and DMSO solution formats for flexible dosing.
  • High purity suitable for biochemical and cellular assays.