Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-(1H-indol-3-yl)-N-(2-oxo-1,2-dihydroquinolin-4-yl)acetamide | 792957-74-5 | 98.0% | 354.40 | C23H18N2O2 | 25 MG
SDP

Catalog No. 5000212344
Encompass_Preferred

This small-molecule inhibitor blocks mitochondrial permeability transition pore (mPTP) opening and is intended for in vitro research. It shows inhibitory activity with an IC50 of 2.8 μM and is supplied as a solid, high-purity research reagent.

  • Inhibits mPTP opening (IC50 2.8 μM).
  • For in vitro research use only.
  • Purity 98.0%.
  • Solid, off-white to light yellow.
  • Soluble in DMSO (50 mg/mL); ultrasonic agitation recommended.
  • Molecular weight 354.40; formula C23H18N2O2.
  • CAS number 792957-74-5.
  • Storage: powder -20°C (3 years), 4°C (2 years).

Catalog No. 50-002-12344 Supplier Medchemexpress LLC Supplier No. HY10085225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This small-molecule inhibitor blocks mitochondrial permeability transition pore (mPTP) opening and is intended for in vitro research. It shows inhibitory activity with an IC50 of 2.8 μM and is supplied as a solid, high-purity research reagent.

  • Inhibits mPTP opening (IC50 2.8 μM).
  • For in vitro research use only.
  • Purity 98.0%.
  • Solid, off-white to light yellow.
  • Soluble in DMSO (50 mg/mL); ultrasonic agitation recommended.
  • Molecular weight 354.40; formula C23H18N2O2.
  • CAS number 792957-74-5.
  • Storage: powder -20°C (3 years), 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.