Learn More
Sigma Aldrich 2-(1H-indol-3-ylsulfanyl)-ethylamine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Reactant for:
- Preparation of potential antibacterial agents
- Asymmetric addition reactions
- Solid-phase synthesis of bodipy dyes and development of an ig fluorescent sensor
- Condensation reactions with phenylenediamine or aminothiophenol catalyzed by mesoporous mixed metal oxide nanocrystals preparaed from aero gel process
Specifications
Specifications
| Molecular Formula | C10H12N2S |
| Linear Formula | C10H12N2S |
| MDL Number | MFCD00297155 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 192.28 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.