Learn More
Medchemexpress LLC 2(1H)-Isoquinolineacetamide, 3,4-dihydro-6,7-dimethoxy-N-methyl-a-phenyl-1-[2-[4-(trifluoromethy | 913358-93-7 | MFCD22419385 | 100.0% | 549.02 g/mol | C29H32ClF3N2O3 | 25 MG
SDP
Catalog No. 5000243042
Encompass_Preferred
Almorexant hydrochloride is the hydrochloride salt of a potent, orally active dual orexin (OX1/OX2) receptor antagonist used in neuroscience research of sleep and arousal mechanisms. It is supplied as a high-purity solid and as a DMSO solution, with batch COA documentation for research use.
  • Potent dual orexin receptor antagonist suitable for receptor pharmacology studies
  • High purity suitable for in vitro and in vivo experiments
  • Available in multiple solid pack sizes and as a DMSO solution
  • Certificate of analysis provided for batch-specific quality assurance
  • Hydrochloride salt form for improved handling and formulation stability
Catalog No. 50-002-43042 Supplier Medchemexpress LLC Supplier No. HY10805A25MG
Almorexant hydrochloride is the hydrochloride salt of a potent, orally active dual orexin (OX1/OX2) receptor antagonist used in neuroscience research of sleep and arousal mechanisms. It is supplied as a high-purity solid and as a DMSO solution, with batch COA documentation for research use.
  • Potent dual orexin receptor antagonist suitable for receptor pharmacology studies
  • High purity suitable for in vitro and in vivo experiments
  • Available in multiple solid pack sizes and as a DMSO solution
  • Certificate of analysis provided for batch-specific quality assurance
  • Hydrochloride salt form for improved handling and formulation stability