Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(1H-Pyrazol-5-yl)aniline 97.0+%, TCI America™
SDP

Catalog No. P19021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19021G Supplier TCI America Supplier No. P19021G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111562-32-4
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD09028476
InChI Key NRSKRRKWPPDKNI-UHFFFAOYSA-N
Synonym 5-(2-Aminophenyl)pyrazole
PubChem CID 257424
IUPAC Name 2-(1H-pyrazol-5-yl)aniline
SMILES C1=CC=C(C(=C1)C2=CC=NN2)N

Specifications

Melting Point 129°C
Quantity 1 g
Formula Weight 159.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(1H-Pyrazol-5-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.