Learn More
Medchemexpress LLC 2(1H)-pyridinone, 3,5,6-trichloro- | 6515-38-4 | MFCD00130269, MFCD02313933 | 99.9% | 198.43 g/mol | C5H2Cl3NO | 1 G
SDP
Catalog No. 5000299902
Encompass_Preferred
3,5,6-Trichloro-2-pyridinol (TCPy) is the primary metabolite of several organophosphate pesticides and is supplied as a research-grade standard for analytical, metabolite, and toxicology studies. It is used as an analytical reference and for metabolism and exposure assessment research.
  • High purity (reported 99.85%) suitable for analytical applications.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Supplied with certificate of analysis and data sheet for quality assurance.
  • Suitable for use as an analytical standard, metabolite reference, and in toxicology studies.
  • Multiple pack sizes available to support small-scale and preparative workflows.
Catalog No. 50-002-99902 Supplier Medchemexpress LLC Supplier No. HYW0181711G
3,5,6-Trichloro-2-pyridinol (TCPy) is the primary metabolite of several organophosphate pesticides and is supplied as a research-grade standard for analytical, metabolite, and toxicology studies. It is used as an analytical reference and for metabolism and exposure assessment research.
  • High purity (reported 99.85%) suitable for analytical applications.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Supplied with certificate of analysis and data sheet for quality assurance.
  • Suitable for use as an analytical standard, metabolite reference, and in toxicology studies.
  • Multiple pack sizes available to support small-scale and preparative workflows.