Learn More
Medchemexpress LLC 2(1H)-pyridinone, 5,6-dihydro-4-(4-methylphenyl)-3-(2H-tetrazol-5-yl)-6-(trifluoromethyl)-(6S) | 1441057-15-3 | 99.0% | 525.45 g·mol⁻¹ | C24H21F6N5O2 | 5 MG
SDP
Catalog No. 5000262615
Encompass_Preferred
BMS-963272 is an orally active, selective inhibitor of monoacylglycerol acyltransferase 2 (MGAT2) used in preclinical and early clinical research to investigate metabolic disorders, liver inflammation, and fibrosis. It shows potent in vitro activity and documented physicochemical and storage properties suitable for research use.
  • Potent MGAT2 inhibition (IC50 7.1 nM).
  • Orally active compound suitable for in vivo studies.
  • High reported purity supports experimental reproducibility.
  • Soluble in DMSO for in vitro assay preparation.
  • Characterized molecular weight and formula for accurate dosing.
  • Documented storage conditions prolong sample stability.
Catalog No. 50-002-62615 Supplier Medchemexpress LLC Supplier No. HY1329245MG
BMS-963272 is an orally active, selective inhibitor of monoacylglycerol acyltransferase 2 (MGAT2) used in preclinical and early clinical research to investigate metabolic disorders, liver inflammation, and fibrosis. It shows potent in vitro activity and documented physicochemical and storage properties suitable for research use.
  • Potent MGAT2 inhibition (IC50 7.1 nM).
  • Orally active compound suitable for in vivo studies.
  • High reported purity supports experimental reproducibility.
  • Soluble in DMSO for in vitro assay preparation.
  • Characterized molecular weight and formula for accurate dosing.
  • Documented storage conditions prolong sample stability.