Learn More
Medchemexpress LLC 2(1H)-pyrimidinone, tetrahydro-1,3-dimethyl- | 7226-23-5 | MFCD00006550 | 99.9% | 128.17 g/mol | C6H12N2O | 100g
SDP
Catalog No. 5000268178
Encompass_Preferred
N N -Dimethyltrimethyleneurea is an intermediate N N -Dimethyltrimethyleneurea can be used in the preparation of naphthalocyanine compounds (near-infrared absorbing dyes)[1]
Catalog No. 50-002-68178 Supplier Medchemexpress LLC Supplier No. HYY0100100G
N N -Dimethyltrimethyleneurea is an intermediate N N -Dimethyltrimethyleneurea can be used in the preparation of naphthalocyanine compounds (near-infrared absorbing dyes)[1]