Learn More
Medchemexpress LLC 2(1H)-Quinolinone, 6-[[3-fluoro-5-(tetrahydro-4-methoxy-2H-pyran-4-yl)phenoxy]methyl]-1-methyl- | 140841-32-3 | 99.9% | 397.44 | 1 MG
SDP
Catalog No. 5000365551
Encompass_Preferred
ZD 2138 is a selective 5-lipoxygenase inhibitor, suitable for research in areas such as asthma. This product is provided as an off-white to light yellow solid with a high purity.
  • High purity: 99.9% (LCMS)
  • Off-white to light yellow solid appearance
  • Consistent with chemical structure (1H NMR, LCMS)
  • Suitable for research use
Catalog No. 50-003-65551 Supplier Medchemexpress LLC Supplier No. 1MGHY169138
ZD 2138 is a selective 5-lipoxygenase inhibitor, suitable for research in areas such as asthma. This product is provided as an off-white to light yellow solid with a high purity.
  • High purity: 99.9% (LCMS)
  • Off-white to light yellow solid appearance
  • Consistent with chemical structure (1H NMR, LCMS)
  • Suitable for research use