Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2(1H)-Quinolinone, 6-[[3-fluoro-5-(tetrahydro-4-methoxy-2H-pyran-4-yl)phenoxy]methyl]-1-methyl- | 140841-32-3 | 99.9% | 397.44 | 25 MG
SDP

Catalog No. 5000366030
Encompass_Preferred

ZD 2138 is a chemical compound identified by CAS No. 140841-32-3. It has a molecular formula of C23H24FNO4 and a molecular weight of 397.44. This compound is typically an off-white to light yellow solid with a high purity of 99.9%.

  • Chemical name: 2(1H)-Quinolinone, 6-[[3-fluoro-5-(tetrahydro-4-methoxy-2H-pyran-4-yl)phenoxy]methyl]-1-methyl-
  • Molecular formula: C23H24FNO4
  • Molecular weight: 397.44
  • Purity: 99.9%
  • Appearance: Off-white to light yellow solid
  • Storage (powder): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-66030 Supplier Medchemexpress LLC Supplier No. 25MGHY169138
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZD 2138 is a chemical compound identified by CAS No. 140841-32-3. It has a molecular formula of C23H24FNO4 and a molecular weight of 397.44. This compound is typically an off-white to light yellow solid with a high purity of 99.9%.

  • Chemical name: 2(1H)-Quinolinone, 6-[[3-fluoro-5-(tetrahydro-4-methoxy-2H-pyran-4-yl)phenoxy]methyl]-1-methyl-
  • Molecular formula: C23H24FNO4
  • Molecular weight: 397.44
  • Purity: 99.9%
  • Appearance: Off-white to light yellow solid
  • Storage (powder): -20°C for 3 years; 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.