Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2(1H)-Quinolinone, 6-[[3-fluoro-5-(tetrahydro-4-methoxy-2H-pyran-4-yl)phenoxy]methyl]-1-methyl- | 140841-32-3 | 99.9% | 397.44 | 5 MG
SDP

Catalog No. 5000364221
Encompass_Preferred

ZD 2138 is an off-white to light yellow solid. This product has been tested and complies with the given specifications.

  • Molecular formula: C23H24FNO4
  • Molecular weight: 397.44
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-64221 Supplier Medchemexpress LLC Supplier No. 5MGHY169138
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZD 2138 is an off-white to light yellow solid. This product has been tested and complies with the given specifications.

  • Molecular formula: C23H24FNO4
  • Molecular weight: 397.44
  • Storage for powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.