Learn More
Medchemexpress LLC 2(1H)-Quinolinone, 7-amino-4-methyl- | 19840-99-4 | 174.20 | 250 MG
SDP
Catalog No. 5000449875
Encompass_Preferred
Carbostyril 124 is a sensitising chromophore that acts as a reasonably effective organic sensitizer for a proximate bound terbium ion. It is exclusively for research use.
  • Sensitising chromophore
  • Functions as an organic sensitizer for terbium ions
  • Exhibits metal-based luminescence with europium and terbium complexes
  • Intended for research use only
Catalog No. 50-004-49875 Supplier Medchemexpress LLC Supplier No. 250MGHYW008292
Carbostyril 124 is a sensitising chromophore that acts as a reasonably effective organic sensitizer for a proximate bound terbium ion. It is exclusively for research use.
  • Sensitising chromophore
  • Functions as an organic sensitizer for terbium ions
  • Exhibits metal-based luminescence with europium and terbium complexes
  • Intended for research use only