Learn More
Ace Glass, Inc. Heating tape, 5/8in, 7ft length, 105w at 115v max, Glas-Col;103A FET0.67
SDP
Catalog No. 50977376
Encompass
5/8 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 7 foot length. Can be operated directly through a temperature control at normal outlet voltage. 105w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.
Catalog No. 50-977-376 Supplier Ace Glass, Inc. Supplier No. 1206306
5/8 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 7 foot length. Can be operated directly through a temperature control at normal outlet voltage. 105w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.