Learn More
Medchemexpress LLC 2-[(1R)-1-[2-[4-methoxy-3-(2-morpholin-4-ylethoxy)phenyl]-5-methyl-1,3-thiazol-4-yl]ethyl]sulfanyl | 2107280-55-5 | MFCD34469884 | 99.5% | C23H30N6O3S2 | 10MG
SDP
Catalog No. 5000258317
Encompass_Preferred
DI-87 is an orally active, selective deoxycytidine kinase (dCK) inhibitor developed for anticancer research. It exhibits potent biochemical activity in the low-nanomolar range and has demonstrated antitumor effects in preclinical models, where it is used to enhance the efficacy of nucleoside analog therapies.
  • Potent dCK inhibition with low-nanomolar EC50.
  • Orally bioavailable and metabolically stable for in vivo studies.
  • Demonstrated antitumor activity in preclinical tumor models.
  • Enhances efficacy of nucleoside analog therapies in combination studies.
  • Supplied as a stable powder, soluble in DMSO for assay preparation.
Catalog No. 50-002-58317 Supplier Medchemexpress LLC Supplier No. HY13314110MG
DI-87 is an orally active, selective deoxycytidine kinase (dCK) inhibitor developed for anticancer research. It exhibits potent biochemical activity in the low-nanomolar range and has demonstrated antitumor effects in preclinical models, where it is used to enhance the efficacy of nucleoside analog therapies.
  • Potent dCK inhibition with low-nanomolar EC50.
  • Orally bioavailable and metabolically stable for in vivo studies.
  • Demonstrated antitumor activity in preclinical tumor models.
  • Enhances efficacy of nucleoside analog therapies in combination studies.
  • Supplied as a stable powder, soluble in DMSO for assay preparation.