Learn More
Medchemexpress LLC 2-((1R,5S)-8-(2-(((S)-4-(3-(N,N-dimethylsulfamoyl)-1H-pyrazole-1-carbonyl)-3-methylpiperazin-1-yl)methyl)-5-... | 3007772-60-0 | 98.1% | C27H36F3N7O5S | 10MG
SDP
Catalog No. 5000260919
Encompass_Preferred
ABD957 is a potent, selective covalent inhibitor of the ABHD17 family of depalmitoylases used as a research reagent to study protein depalmitoylation and NRAS signaling in biochemical and cell-based assays. It exhibits an IC50 of 0.21 μM against ABHD17B and is supplied as a white to off-white solid.
  • Potent, selective covalent inhibition of ABHD17 enzymes (IC50 0.21 μM for ABHD17B).
  • Suitable for biochemical and cell-based studies of depalmitoylation and signaling.
  • High purity (98.1%) for reliable experimental results.
  • Available in multiple small-scale pack sizes for research use.
  • Stable when stored under recommended conditions for long-term use.
Catalog No. 50-002-60919 Supplier Medchemexpress LLC Supplier No. HY14216110MG
ABD957 is a potent, selective covalent inhibitor of the ABHD17 family of depalmitoylases used as a research reagent to study protein depalmitoylation and NRAS signaling in biochemical and cell-based assays. It exhibits an IC50 of 0.21 μM against ABHD17B and is supplied as a white to off-white solid.
  • Potent, selective covalent inhibition of ABHD17 enzymes (IC50 0.21 μM for ABHD17B).
  • Suitable for biochemical and cell-based studies of depalmitoylation and signaling.
  • High purity (98.1%) for reliable experimental results.
  • Available in multiple small-scale pack sizes for research use.
  • Stable when stored under recommended conditions for long-term use.