Learn More
Medchemexpress LLC 2,2'-[1,2-phenylenebis(oxy)]bis[N-phenyl-N-(phenylmethyl)acetamide] | 61595-77-5 | >98.0% | 556.65 | C36H32N2O4 | 5 MG
SDP
Catalog No. 5000222615
Encompass_Preferred
ETH 157 (Sodium ionophore II) is a neutral sodium (Na+) ionophore used in research to induce Na+/Ca2+ selectivity in membranes. It is employed in analytical chemistry and membrane electrode applications to selectively transport or detect sodium ions. Chemical formula C36H32N2O4; molecular weight 556.65; supplied as a solid.
Catalog No. 50-002-22615 Supplier Medchemexpress LLC Supplier No. HY1371725MG
ETH 157 (Sodium ionophore II) is a neutral sodium (Na+) ionophore used in research to induce Na+/Ca2+ selectivity in membranes. It is employed in analytical chemistry and membrane electrode applications to selectively transport or detect sodium ions. Chemical formula C36H32N2O4; molecular weight 556.65; supplied as a solid.