Learn More
Medchemexpress LLC 2,2'-(1,2-phenylenebis(oxy))bis(N-phenyl-N-(phenylmethyl)acetamide) | 61595-77-5 | MFCD00042979 | >98.0% | C36H32N2O4 | 10MG
SDP
Catalog No. 5000259340
Encompass_Preferred
ETH 157 (CAS 61595-77-5) is a sodium ionophore that induces Na+/Ca2+ selectivity in membranes. Supplied as a solid research chemical, it is intended for experimental modulation of sodium transport across lipid bilayers and other membrane studies. It exhibits high solubility in DMSO and has recommended cold storage for long-term stability.
  • Sodium-selective ionophore for membrane transport studies.
  • Induces Na+/Ca2+ selectivity in lipid membranes.
  • High solubility in DMSO (100 mg/mL) with ultrasonic assistance.
  • Stable as a powder when stored cold (powder: -20°C for long-term storage).
  • Supplied in small research quantities suitable for laboratory use.
Catalog No. 50-002-59340 Supplier Medchemexpress LLC Supplier No. HY13717210MG
ETH 157 (CAS 61595-77-5) is a sodium ionophore that induces Na+/Ca2+ selectivity in membranes. Supplied as a solid research chemical, it is intended for experimental modulation of sodium transport across lipid bilayers and other membrane studies. It exhibits high solubility in DMSO and has recommended cold storage for long-term stability.
  • Sodium-selective ionophore for membrane transport studies.
  • Induces Na+/Ca2+ selectivity in lipid membranes.
  • High solubility in DMSO (100 mg/mL) with ultrasonic assistance.
  • Stable as a powder when stored cold (powder: -20°C for long-term storage).
  • Supplied in small research quantities suitable for laboratory use.