Learn More
Medchemexpress LLC 2,2'-(1,4-phenylene)bis(4,5-diphenyl-1H-imidazole) | 4051-59-6 | 98.0% | 514.62 g/mol | C36H26N4 | 10 MG
SDP
Catalog No. 5000247410
Encompass_Preferred
MSX-130 is a small-molecule antagonist of the C-X-C chemokine receptor 4 (CXCR4) used in research to inhibit CXCR4-mediated signaling and reduce cancer metastasis. It is supplied as a solid research reagent with defined storage and solubility recommendations suitable for in vitro and in vivo studies.
  • Small-molecule CXCR4 antagonist for research applications.
  • High purity (98.0%) suitable for biochemical assays.
  • Soluble in DMSO (≈2 mg/mL with warming or sonication).
  • Stable as powder when stored at recommended temperatures.
  • Available in common research package sizes for dose-response work.
Catalog No. 50-002-47410 Supplier Medchemexpress LLC Supplier No. HY10301010MG
MSX-130 is a small-molecule antagonist of the C-X-C chemokine receptor 4 (CXCR4) used in research to inhibit CXCR4-mediated signaling and reduce cancer metastasis. It is supplied as a solid research reagent with defined storage and solubility recommendations suitable for in vitro and in vivo studies.
  • Small-molecule CXCR4 antagonist for research applications.
  • High purity (98.0%) suitable for biochemical assays.
  • Soluble in DMSO (≈2 mg/mL with warming or sonication).
  • Stable as powder when stored at recommended temperatures.
  • Available in common research package sizes for dose-response work.