Learn More
Medchemexpress LLC 2-[2-[2-[2-(2-Bromoethoxy)ethoxy]ethoxy]ethoxy]oxane | 1803547-60-5 | 97.0% | 341.24 | 250 MG
SDP
Catalog No. 5000348251
Encompass_Preferred
Br-PEG4-THP is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-48251 Supplier Medchemexpress LLC Supplier No. 250MGHY138372
Br-PEG4-THP is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker; one targets an E3 ubiquitin ligase and the other targets a specific protein. This mechanism leverages the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits ubiquitin-proteasome system
  • Selectively degrades target proteins