Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-(2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluorooctane-1,8-diyl)bis(oxirane), TCI America™
SDP

Catalog No. D49915G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49915G 5 g
D499125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49915G Supplier TCI America Supplier No. D49915G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74328-56-6
Molecular Formula C12H10F12O2
Molecular Weight (g/mol) 414.191
InChI Key BUCVATDLDBZNPM-UHFFFAOYSA-N
PubChem CID 15507707
IUPAC Name 2-[2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-8-(oxiran-2-yl)octyl]oxirane
SMILES C1C(O1)CC(C(C(C(C(C(CC2CO2)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 106°C
Quantity 5 g
Formula Weight 414.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2′-(2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluorooctane-1,8-diyl)bis(oxirane)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.